Anti-CCBE1 Rabbit Polyclonal Antibody
Catalog # 10581-344
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Collagen And Calcium binding EGF Domains 1
- Antigen Symbol:CCBE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:147372
- Antigen Synonyms:CCBE1_HUMAN|CCBE 1|Collagen and calcium-binding EGF domain-containing protein 1|Collagen and Calcium binding EGF domain containing protein 1|ccbe1|Collagen and Calcium binding EGF domains 1|Full of fluid protein homolog.
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10581-344
- Supplier no.:H00147372-D01P
Specifications
About this item
Type: Primary
Antigen: CCBE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human