Order Entry
United States
ContactUsLinkComponent
 
Anti-MESP2 Mouse Monoclonal Antibody [clone: 1C8]
  10593-254
 :  Abnova
undefined
Anti-MESP2 Mouse Monoclonal Antibody [clone: 1C8]
  10593-254
 :  Abnova
 :  H00145873-M01

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Mesoderm posterior 2 homolog
  • Antigen Symbol:
    MESP2
  • Clonality:
    Monoclonal
  • Clone:
    1C8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Gene ID:
    145873
  • Antigen Synonyms:
    bHLHc6|SCDO2
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    10593-254
  • Supplier no.:
    H00145873-M01

 

 

Type: Primary
Antigen: MESP2
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human