Order Entry
United States
ContactUsLinkComponent
Anti-ACCS Rabbit Polyclonal Antibody
Anti-ACCS Rabbit Polyclonal Antibody
  10596-808
 :  Abnova
undefined
Anti-ACCS Rabbit Polyclonal Antibody
  10596-808
 :  Abnova
 :  H00084680-D01P

 

  • Antibody Type:
    Primary
  • Antigen Name:
    1-aminocyclopropane-1-carboxylate synthase homolog (Arabidopsis)(non-functional)
  • Antigen Symbol:
    ACCS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Gene ID:
    84680
  • Antigen Synonyms:
    ACS|PHACS
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    10596-808
  • Supplier no.:
    H00084680-D01P

 

 

Type: Primary
Antigen: ACCS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human