Order Entry
United States
ContactUsLinkComponent
Anti-TRIM63 Mouse Monoclonal Antibody [clone: 6G6]
Anti-TRIM63 Mouse Monoclonal Antibody [clone: 6G6]
Catalog # 10625-476
Supplier:  Abnova
undefined
Anti-TRIM63 Mouse Monoclonal Antibody [clone: 6G6]
Catalog # 10625-476
Supplier:  Abnova
Supplier Number:  H00084676-M01A

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Tripartite Motif Containing 63
  • Antigen Symbol:
    TRIM63
  • Clonality:
    Monoclonal
  • Clone:
    6G6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    200 µL
  • Cross Adsorption:
    No
  • Gene ID:
    84676
  • Antigen Synonyms:
    tripartite motif-containing 63|muscle specific ring finger protein 2|MURF1|IRF|tripartite motif-containing protein 63|SMRZ|RNF28|iris ring finger protein|striated muscle RING zinc finger protein|MURF2|E3 ubiquitin-protein ligase TRIM63|muscle-specific RING finger protein 1|ring finger protein 28
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    10625-476
  • Supplier no.:
    H00084676-M01A

Specifications

About this item

Type: Primary
Antigen: TRIM63
Clonality: Monoclonal
Clone: 6G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human