Anti-FRMD8 Mouse Polyclonal Antibody
Catalog # 10556-660
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:FERM Domain Containing 8
- Antigen Symbol:FRMD8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:83786
- Antigen Synonyms:FKSG44
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10556-660
- Supplier no.:H00083786-B01P
Specifications
About this item
Type: Primary
Antigen: FRMD8 (FERM domain containing 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse