Anti-NDST4 Mouse Polyclonal Antibody
Catalog # 10564-286
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:N-Deacetylase/N-Sulfotransferase (Heparan Glucosaminyl) 4
- Antigen Symbol:NDST4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:64579
- Antigen Synonyms:NDST-4|NHSST4
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10564-286
- Supplier no.:H00064579-A01
Specifications
About this item
Type: Primary
Antigen: NDST4 (N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human