Anti-ELMO2 Mouse Monoclonal Antibody [clone: 3G7]
Catalog # 10584-932
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:engulfment and cell motility 2
- Antigen Symbol:ELMO2
- Clonality:Monoclonal
- Clone:3G7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Gene ID:63916
- Antigen Synonyms:ELMO-2|RP11-394O2.2|ced-12 homolog 2|KIAA1834|C. elegans)|engulfment and cell motility 2 (ced-12 homolog|PH domain protein CED12A|CED12|CED-12|engulfment and cell motility 2|FLJ11656
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10584-932
- Supplier no.:H00063916-M02
Specifications
About this item
Type: Primary
Antigen: ELMO2
Clonality: Monoclonal
Clone: 3G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human