Anti-DUSP22 Rabbit Polyclonal Antibody
Catalog # 10584-428
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Dual Specificity Phosphatase 22
- Antigen Symbol:DUSP22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Size:100 µL
- Cross Adsorption:No
- Gene ID:56940
- Antigen Synonyms:JSP1|LMWDSP2|MKPX|LMW-DSP2|JKAP|MKP-x|JSP-1|VHX
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10584-428
- Supplier no.:H00056940-D01
Specifications
About this item
Type: Primary
Antigen: DUSP22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human