Anti-PBK Mouse Monoclonal Antibody [clone: 6000]
Catalog # 10596-120
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:PDZ binding kinase
- Antigen Symbol:PBK
- Clonality:Monoclonal
- Clone:6E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Gene ID:55872
- Antigen Synonyms:TOPK|T-lymphokine-activated killer cell-originated protein kinase|MAPKK-like protein kinase|PBK|Nori-3|SPK|Spermatogenesis-related protein kinase|PDZ-binding kinase|T-LAK cell-originated protein kinase
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10596-120
- Supplier no.:H00055872-M27
Specifications
About this item
Type: Primary
Antigen: PBK
Clonality: Monoclonal
Clone: 6000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human