Anti-NUP133 Mouse Monoclonal Antibody [clone: 3E8]
Catalog # 10595-558
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Nucleoporin 133kDa
- Antigen Symbol:NUP133
- Clonality:Monoclonal
- Clone:3E8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:55746
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10595-558
- Supplier no.:H00055746-M01
Specifications
About this item
Type: Primary
Antigen: NUP133
Clonality: Monoclonal
Clone: 3E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human