Order Entry
United States
ContactUsLinkComponent
Anti-ZNF280C Mouse Polyclonal Antibody
Anti-ZNF280C Mouse Polyclonal Antibody
Catalog # 10579-896
Supplier:  Avantor
undefined
Anti-ZNF280C Mouse Polyclonal Antibody
Catalog # 10579-896
Supplier:  Avantor

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Zinc Finger Protein 280C
  • Antigen Symbol:
    ZNF280C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 µL
  • Cross Adsorption:
    No
  • Gene ID:
    55609
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    10579-896
  • Supplier no.:
    H00055609-B01

Specifications

About this item

Type: Primary
Antigen: ZNF280C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human