Anti-CECR1 Mouse Monoclonal Antibody [clone: 3D11]
Catalog # 10581-474
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Cat Eye Syndrome Chromosome Region, Candidate 1
- Antigen Symbol:CECR1
- Clonality:Monoclonal
- Clone:3D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:51816
- Antigen Synonyms:ADA2|SNEDS|IDGFL|ADGF|PAN
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10581-474
- Supplier no.:H00051816-M01
Specifications
About this item
Type: Primary
Antigen: CECR1
Clonality: Monoclonal
Clone: 3D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: