Anti-GOLM1 Mouse Monoclonal Antibody [clone: 5B10]
Catalog # 10587-658
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Golgi Membrane Protein 1
- Antigen Symbol:GOLM1
- Clonality:Monoclonal
- Clone:5B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:51280
- Antigen Synonyms:GP73|FLJ22634|golgi membrane protein GP73|golgi membrane protein 1|C9orf155|GOLPH2|GOLM1|FLJ23608|golgi phosphoprotein 2|bA379P1.3|golgi protein|PSEC0257|RP11-379P1.3|73-kD
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10587-658
- Supplier no.:H00051280-M06
Specifications
About this item
Type: Primary
Antigen: GOLM1 (golgi membrane protein 1)
Clonality: Monoclonal
Clone: 5B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human