Anti-PIPOX Mouse Polyclonal Antibody
Catalog # 10567-598
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Pipecolic Acid Oxidase
- Antigen Symbol:PIPOX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:51268
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10567-598
- Supplier no.:H00051268-B01
Specifications
About this item
Type: Primary
Antigen: PIPOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human