Anti-CIDEB Mouse Monoclonal Antibody [clone: 2A10]
Catalog # 10582-252
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Cell death-inducing DFFA-like effector b
- Antigen Symbol:CIDEB
- Clonality:Monoclonal
- Clone:2A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:27141
- Antigen Synonyms:CIDEB_HUMAN.|Cell death activator CIDE B|Cell death inducing DFFA like effector B|Cell death activator CIDE-B|CIDEB|Cell death-inducing DFFA-like effector B
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10582-252
- Supplier no.:H00027141-M01
Specifications
About this item
Type: Primary
Antigen: CIDEB
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: