Anti-TRAF2 And NCK Interacting Kinase Mouse Monoclonal Antibody [clone: 40000]
Catalog # 10604-306
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Traf2 And Nck Interacting Kinase
- Antigen Symbol:TNIK
- Clonality:Monoclonal
- Clone:4E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:23043
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10604-306
- Supplier no.:H00023043-M02
Specifications
About this item
Type: Primary
Antigen: TRAF2 And NCK Interacting Kinase
Clonality: Monoclonal
Clone: 40000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human