Anti-SLC17A3 Mouse Polyclonal Antibody
Catalog # 10579-214
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 17 Member A3
- Antigen Symbol:SLC17A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:10786
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10579-214
- Supplier no.:H00010786-B01P
Specifications
About this item
Type: Primary
Antigen: SLC17A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human