Anti-TOMM20 (translocase of outer mitochondrial membrane 20 homolog (yeast)) Mouse Monoclonal Antibody [clone: 4F3]
Catalog # 10604-390
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Translocase of Outer Mitochondrial Membrane 20
- Antigen Symbol:TOMM20
- Clonality:Monoclonal
- Clone:4F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:9804
- Antigen Synonyms:KIAA0016
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10604-390
- Supplier no.:H00009804-M01
Specifications
About this item
Type: Primary
Antigen: TOMM20 (translocase of outer mitochondrial membrane 20 homolog (yeast))
Clonality: Monoclonal
Clone: 4F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat