Anti-PAPSS2 Mouse Polyclonal Antibody
10566-592
: Abnova
- Antibody Type:Primary
- Antigen Name:3'-phosphoadenosine 5'-phosphosulfate Synthase 2
- Antigen Symbol:PAPSS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:9060
- Antigen Synonyms:Sulfurylase kinase 2|PAPS synthethase 2|SK2|Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10566-592
- Supplier no.:H00009060-B01P
Type: Primary
Antigen: PAPSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human