Anti-PROZ Rabbit Polyclonal Antibody
Catalog # 10598-234
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Z, vitamin K-dependent plasma glycoprotein
- Antigen Symbol:PROZ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:8858
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10598-234
- Supplier no.:H00008858-D01P
Specifications
About this item
Type: Primary
Antigen: PROZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human