Anti-WRN Mouse Monoclonal Antibody [clone: 2F7]
Catalog # 10605-828
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Werner syndrome, RecQ helicase-like
- Antigen Symbol:WRN
- Clonality:Monoclonal
- Clone:2F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:7486
- Antigen Synonyms:RECQL3|RECQ3|RECQL2
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10605-828
- Supplier no.:H00007486-M06
Specifications
About this item
Type: Primary
Antigen: WRN
Clonality: Monoclonal
Clone: 2F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human