Anti-VDAC1 Mouse Monoclonal Antibody [clone: 2A8]
10625-588
: Abnova
- Antibody Type:Primary
- Antigen Name:Voltage-Dependent Anion Channel 1
- Antigen Symbol:VDAC1
- Clonality:Monoclonal
- Clone:2A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:200 µL
- Cross Adsorption:No
- Gene ID:7416
- Antigen Synonyms:hVDAC1|Plasmalemmal porin|Outer mitochondrial membrane protein porin 1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10625-588
- Supplier no.:H00007416-M28A
Type: Primary
Antigen: VDAC1
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human