Anti-TYR Mouse Polyclonal Antibody
Catalog # 10580-376
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Tyrosinase
- Antigen Symbol:TYR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:7299
- Antigen Synonyms:Tumor rejection antigen AB|ATN|CMM8|SHEP3|Monophenol monooxygenase|OCAIA|OCA1A|OCA1|TYR|LB24-AB|SK29-AB
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10580-376
- Supplier no.:H00007299-B01P
Specifications
About this item
Type: Primary
Antigen: TYR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human