Anti-TTK Mouse Monoclonal Antibody [clone: 2F1]
Catalog # 10604-968
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:TTK Protein kinase
- Antigen Symbol:TTK
- Clonality:Monoclonal
- Clone:2F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:7272
- Antigen Synonyms:CT96|MPH1|ESK|MPS1|PYT|MPS1L1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10604-968
- Supplier no.:H00007272-M04
Specifications
About this item
Type: Primary
Antigen: TTK
Clonality: Monoclonal
Clone: 2F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human