Anti-SCNN1D Rabbit Polyclonal Antibody
Catalog # 10600-758
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Sodium Channel Nonvoltage-gated 1 Delta
- Antigen Symbol:SCNN1D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:6339
- Antigen Synonyms:ENaCd|SCNED|ENaCdelta|dNaCh
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10600-758
- Supplier no.:H00006339-D01P
Specifications
About this item
Type: Primary
Antigen: SCNN1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human