Anti-ACSM3 Rabbit Polyclonal Antibody
10580-808
: Abnova
- Antibody Type:Primary
- Antigen Name:Acyl-CoA Synthetase Medium-chain Family Member 3
- Antigen Symbol:ACSM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:6296
- Antigen Synonyms:SAH|SA
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10580-808
- Supplier no.:H00006296-D01P
Type: Primary
Antigen: ACSM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse