Anti-PROX1 Mouse Monoclonal Antibody [clone: 4D1]
10598-228
: Abnova
- Antibody Type:Primary
- Antigen Name:Prospero Homeobox 1
- Antigen Symbol:PROX1
- Clonality:Monoclonal
- Clone:4D1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:5629
- Antigen Synonyms:PROX-1|prospero-related homeobox 1|prospero homeobox protein 1|homeobox prospero-like protein PROX1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10598-228
- Supplier no.:H00005629-M06
Type: Primary
Antigen: PROX1
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human