Anti-POU6F1 Rabbit Polyclonal Antibody
10597-686
: Abnova
- Antibody Type:Primary
- Antigen Name:POU Domain Class 6, Transcription Factor 1
- Antigen Symbol:POU6F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:5463
- Antigen Synonyms:MPOU|TCFB1|BRN5
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10597-686
- Supplier no.:H00005463-D01P
Type: Primary
Antigen: POU6F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human