Anti-NPTX1 Mouse Monoclonal Antibody [clone: 7E6]
Catalog # 10595-072
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Neuronal Pentraxin 1
- Antigen Symbol:NPTX1
- Clonality:Monoclonal
- Clone:7E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:4884
- Antigen Synonyms:NP1|NP-I|NPTX1|Neuronal pentraxin-1|Neuronal pentraxin I
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10595-072
- Supplier no.:H00004884-M06
Specifications
About this item
Type: Primary
Antigen: NPTX1
Clonality: Monoclonal
Clone: 7000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human