Anti-NEUROD2 Rabbit Polyclonal Antibody
Catalog # 10594-718
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Neurogenic Differentiation 2
- Antigen Symbol:NEUROD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:4761
- Antigen Synonyms:Class A basic helix-loop-helix protein 1|Neuro-D2|NDRF|neurogenic dferentiation 2|NeuroD-related factor|NDR2|neuroD related factor|NeuroD2|NDF2_HUMAN|Neurogenic dferentiation factor 2|class A basic helix loop helix protein 1|neurogenic basic helix loop helix protein|neuronal dferentiation 2.|bHLHa1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10594-718
- Supplier no.:H00004761-D01P
Specifications
About this item
Type: Primary
Antigen: NEUROD2 (neuronal differentiation 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human