Anti-ILK Mouse Monoclonal Antibody [clone: 4F10]
Catalog # 10590-420
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin-Linked Kinase
- Antigen Symbol:ILK
- Clonality:Monoclonal
- Clone:4F10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- ChIP:Yes
- Gene ID:3611
- Antigen Synonyms:ILK1|Integrin-linked protein kinase|ILK2|p59ILK |59 kDa serine/threonine-protein kinase|ILK-1|ILK-2
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10590-420
- Supplier no.:H00003611-M01
Specifications
About this item
Type: Primary
Antigen: ILK
Clonality: Monoclonal
Clone: 4F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat