Anti-TNC Mouse Polyclonal Antibody
50 µL
Catalog # 10572-566
Marketsource
Supplier: Abnova
Supplier #: H00003371-A01
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type: Primary
Antigen: TNC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Documents
Literature reference - PubMed ID 21110520
Additional Product Details
TDS_ABNOH00003371-A01.pdf
Datasheet
Specifications
- Size:50 µL
- Cross Adsorption:No
- Clonality:Polyclonal
- Host:Mouse
- Cat. No.:10572-566
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:Ice
- Antibody Type:Primary
- Antigen Symbol:TNC
- Gene ID:3371
- Conjugation:Unconjugated
- Antigen Synonyms:HXB|JI|TN|GMEM|150-225|GP|DFNA56|TN-C
- Antigen Name:Tenascin C
- ELISA:Yes
- Storage Temperature:Cool
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



