Anti-HSPA9 Rabbit Polyclonal Antibody
Catalog # 10589-486
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Heat Shock 70 kDa Protein 9 (mortalin)
- Antigen Symbol:HSPA9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:3313
- Antigen Synonyms:MOT|GRP-75|CRP40|GRP75|MOT2|HSPA9B|CSA|HEL-S-124m|PBP74|MTHSP75
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10589-486
- Supplier no.:H00003313-D01P
Specifications
About this item
Type: Primary
Antigen: HSPA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human