Anti-FOXA3 Mouse Polyclonal Antibody
Catalog # 10556-998
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Forkhead Box A3
- Antigen Symbol:FOXA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:3171
- Antigen Synonyms:TCF3G|HNF3G|FKHH3
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10556-998
- Supplier no.:H00003171-B01P
Specifications
About this item
Type: Primary
Antigen: FOXA3 (forkhead box A3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human