Anti-HAL Mouse Monoclonal Antibody [clone: 4F2]
10588-282
: Abnova
- Antibody Type:Primary
- Antigen Name:histidine ammonia-lyase
- Antigen Symbol:HAL
- Clonality:Monoclonal
- Clone:4F2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:3034
- Antigen Synonyms:HSTD|HIS
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10588-282
- Supplier no.:H00003034-M04
Type: Primary
Antigen: HAL (histidine ammonia-lyase)
Clonality: Monoclonal
Clone: 4F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human