Anti-RAPGEF1 Mouse Monoclonal Antibody [clone: 3D10]
Catalog # 10599-158
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Rap Guanine Nucleotide Exchange Factor 1
- Antigen Symbol:RAPGEF1
- Clonality:Monoclonal
- Clone:3D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:2889
- Antigen Synonyms:GRF2|C3G
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10599-158
- Supplier no.:H00002889-M01
Specifications
About this item
Type: Primary
Antigen: RAPGEF1
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human