Anti-ENO2 Mouse Monoclonal Antibody [clone: 1A3]
Catalog # 10584-606
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Enolase 2 (gamma, neuronal)
- Antigen Symbol:ENO2
- Clonality:Monoclonal
- Clone:1A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:2026
- Antigen Synonyms:Gamma-enolase|Enolase 2|ENO2|Neural enolase|ENO-2|2-phospho-D-glycerate hydro-lyase|Neuron-specific enolase
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10584-606
- Supplier no.:H00002026-M01
Specifications
About this item
Type: Primary
Antigen: ENO2
Clonality: Monoclonal
Clone: 1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human