Anti-E2F4 Rabbit Polyclonal Antibody
Catalog # 10584-522
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:E2F transcription factor 4, p107/p130-binding
- Antigen Symbol:E2F4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:1874
- Antigen Synonyms:E2F-4
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10584-522
- Supplier no.:H00001874-D01P
Specifications
About this item
Type: Primary
Antigen: E2F4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse