Anti-CHEK1 Mouse Monoclonal Antibody [clone: 2G3]
Catalog # 10582-120
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Checkpoint Kinase 1
- Antigen Symbol:CHEK1
- Clonality:Monoclonal
- Clone:2G3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Gene ID:1111
- Antigen Synonyms:kinase Chk1|CHK1|EC 2.7.11.1|CHEK1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10582-120
- Supplier no.:H00001111-M05
Specifications
About this item
Type: Primary
Antigen: CHEK1
Clonality: Monoclonal
Clone: 2G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: