Anti-CDH1 Mouse Monoclonal Antibody [clone: 3F4]
Catalog # 10581-680
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Cadherin 1
- Antigen Symbol:CDH1
- Clonality:Monoclonal
- Clone:3F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:999
- Antigen Synonyms:CD324|epithelial Cadherin|Cadherin-E|ECAD|Arc-1|CAM 120/80|cell-CAM 120/80|E-Cadherin|uvomorulin|Cadherin 1|Cadherin-1|E-Cadherin epithelial|calcium-dependent adhesion protein|epithelial|LCAM|CDHE|UVO
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10581-680
- Supplier no.:H00000999-M01
Specifications
About this item
Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: