Anti-MPPED2 Mouse Polyclonal Antibody
Catalog # 10564-620
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:Metallophosphoesterase Domain Containing 2
- Antigen Symbol:MPPED2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:744
- Antigen Synonyms:dJ873F21.1|FAM 1B|Fetal brain protein 239|Chromosome 11 open reading frame 8|Metallophosphoesterase domain containing protein 2|Metallophosphoesterase domain containing 2|dJ1024C24.1|FAM1B|C11orf8|MPPD2_HUMAN.|Hs.46638|D11S302E|239FB|MPPED 2|Brain protein 239
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10564-620
- Supplier no.:H00000744-B01P
Specifications
About this item
Type: Primary
Antigen: MPPED2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat