Anti-BID Mouse Monoclonal Antibody [clone: 3E8-1B10]
Catalog # 10577-900
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:BH3 interacting domain death agonist
- Antigen Symbol:BID
- Clonality:Monoclonal
- Clone:3E8-1B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Gene ID:637
- Antigen Synonyms:MGC15319|MGC42355|FP497
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10577-900
- Supplier no.:H00000637-M03
Specifications
About this item
Type: Primary
Antigen: BID
Clonality: Monoclonal
Clone: 3E8-1B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human