Anti-ATOH1 Mouse Monoclonal Antibody [clone: 1F3]
Catalog # 10577-464
Supplier: Abnova
Specifications
- Antibody Type:Primary
- Antigen Name:atonal bHLH transcription factor 1
- Antigen Symbol:ATOH1
- Clonality:Monoclonal
- Clone:1F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- Gene ID:474
- Antigen Synonyms:bHLHa14|ATH1|HATH1|MATH-1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10577-464
- Supplier no.:H00000474-M03
Specifications
About this item
Type: Primary
Antigen: ATOH1
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human