Anti-ARL1 Mouse Polyclonal Antibody
10559-306
: Abnova
- Antibody Type:Primary
- Antigen Name:ADP-Ribosylation Factor-Like 1
- Antigen Symbol:ARL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Cross Adsorption:No
- Gene ID:400
- Antigen Synonyms:ADP-ribosylation factor-like protein 1|ARFL1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10559-306
- Supplier no.:H00000400-A01
Type: Primary
Antigen: ARL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human