ContactUsLinkComponent
Gravimetric Analysis
Gravimetric Analysis
Catalog # 470163-120
Supplier:  Ward's Science
undefined
Gravimetric Analysis
Catalog # 470163-120
Supplier:  Ward's Science
Supplier Number:  IS8028
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Gravimetric Analysis
  • No. of lab groups:
    15
  • Cat. No.:
    470163-120
  • Includes:
    Teacher's Guide and Student Copymaster
  • Supplier no.:
    IS8028
Specifications
About this item
Determine the Unknown!
  • Classic AP Chemistry Experiment
  • Instructions Included
Determine the identity of a metal carbonate by gravimetric analysis. Gravimetric analysis is a technique through which the amount of an analyte (the ion being analyzed) can be determined through the measurement of mass. You will complete the analyses comparing the masses of two compounds containing the analyte. The formula weight and the identity of the unknown are determined using gravimetric analysis.
Includes:
Calcium Chloride, anhydrous
Silver Nitrate