ContactUsLinkComponent
Determination of the Hardness of Water
  470163-106
undefined
Determination of the Hardness of Water
  470163-106
 :  IS8020
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Determination of the Hardness of Water
  • No. of lab groups:
    15
  • Cat. No.:
    470163-106
  • Includes:
    Teacher's Guide and Student Copymaster
  • Supplier no.:
    IS8020
What Ions Are Present In Water?
  • Classic AP Chemistry Experiment
  • Instructions Included
In this experiment the indicator eriochrome black T (EBT) is used to signal the presence of ions in the water sample. EBT binds with free metal ions in the water to form a pink complex. EDTA has a stronger affinity for the metal ions than EBT so when EDTA is added it replaces the EBT and the EBT returns to its blue, uncomplexed color. The blue color is used as the end point in the titration. A sample of tap water is treated with EBT indicator. If the indicator turns from blue to pink, metal ions such as calcium and magnesium are present. To determine the concentration of ions present, the sample is titrated with a known molar concentration of EDTA.
Includes:
EDTA Solution
Buffer Solution
EBT Solution (Eriochrome Black)