Specifications
- Description:Normal Rabbit Serum
- Size:20 mL
- Environmentally Preferable:
- Cat. No.:101098-390
- Supplier no.:S-5000
Specifications
About this item
Normal Rabbit serum can be used as a blocking reagent in Immunoassays.
- Pooled sample
- Collected from healthy adult animals
- Heat-treated and centrifuged
It is a pooled sample collected from healthy adult animals. The serum is heat-treated and centrifuged to remove precipitates and then filtered. Each serum is tested with the appropriate antibody to check for possible cross-reactivities. The sera are supplied undiluted with 0.08% sodium azide as a preservative.