ContactUsLinkComponent
Goat Serum, Vector Laboratories
Goat Serum, Vector Laboratories
Catalog # 101098-382
undefined
Goat Serum, Vector Laboratories
Catalog # 101098-382
Supplier Number:  S-1000-20
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Normal Whole Goat Serum
  • Size:
    20 mL
  • Cat. No.:
    101098-382
  • Supplier no.:
    S-1000
Specifications
About this item
Normal Goat serum can be used as a blocking reagent in Immunoassays.
  • Can be used for blocking non-specific binding or as an antibody diluent
  • Pooled samples collected from healthy adult animals
  • Tested with appropriate antibody to check for possible cross-reactivities
The serum is heat-treated and centrifuged to remove precipitates and then filtered. The sera are supplied undiluted with 0.08% sodium azide as a preservative.