Order Entry
United States
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # PAW4021
undefined
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # PAW4021
Supplier Number:  W4021
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    300 μl
  • Epitope:
    Heavy and Light chain
  • Storage Temperature:
    2 °C to 10 °C
  • Concentration:
    1 mg/ml
  • Tested Applications:
    Dot Blot
  • Purification:
    affinity-purified
  • Cat. No.:
    PAW4021
  • Supplier no.:
    W4021

Specifications

About this item

Polyclonal secondary antibodies raised in goat, immunoaffinity-purified using immobilized antigens and conjugated to horseradish peroxidase (HRP) enzyme. The mouse antibodies react with all IgG subclasses.

  • Secondary Antibody That Binds to Both Heavy and Light Chains for All Mouse IgG Subclasses
  • Extensively used and published in numerous research articles
  • Antibody is ready-to-use

Polyclonal secondary antibodies raised in goat, immunoaffinity-purified using immobilized antigens and conjugated to horseradish peroxidase (HRP) enzyme. The Anti-Mouse IgG (H+L), HRP Conjugate, antibody binds to both heavy and light chains for all IgG subclasses. As with all antibodies, in certain applications some species-dependent antigen-dependent cross-reactivity may be observed. The product (unless otherwise noted) is supplied as a 1mg/ml solution. A starting working dilution of 1:2,500 is suggested for most Western blot, dot blot and ELISA applications. The optimum concentration of secondary antibody depends on the application and will need to be empirically determined.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse