ContactUsLinkComponent
Dissociation Constants of Weak Acids Kit
Dissociation Constants of Weak Acids Kit
Catalog # 470163-082
Supplier:  Ward's Science
undefined
Dissociation Constants of Weak Acids Kit
Catalog # 470163-082
Supplier:  Ward's Science
Supplier Number:  IS8008
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Dissociation Constants of Weak Acids Kit
  • No. of lab groups:
    15
  • Cat. No.:
    470163-082
  • Supplier no.:
    IS8008
Specifications
About this item
Its All About Equilibrium!
  • Classic AP Chemistry Experiment
  • Instructions Included
Students determine the equilibrium constant of three unknown acids and compare the data to known values to identify them.
Ordering information: Includes Benzoic Acid Crystals, Potassium Hydrogen Phthalate, Sodium Bisulfate, Sodium Hydroxide, and Phenolphthalein Solution.